Kluczowe dokumenty
Z544825
Sigma-Aldrich
Aldrich® high-efficiency variable take off head
joint: ST/NS 29/32, thermometer joint: ST/NS 10/18
About This Item
Kod UNSPSC:
41104812
NACRES:
NB.43
Połączenie
thermometer joint: ST/NS10/18
joint: ST/NS29/32
Powiązane kategorie
Opis ogólny
Vapor from the distillation column passes through the upper holes in the take-off head and into the condenser. The condensed liquid can be drained into a side storage flask (PTFE valve open), or allowed to return to the distillation column (PTFE valve closed) through a slot in the lower part of the head. The upper and lower ST joints permit the use of take-off head with different distillation columns and condensers. Features a precision PTFE valve, and vacuum/purge hose connections with a PTFE stopcock.
A 50mm immersion length thermometer is recommended.
Informacje prawne
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Odwiedź Bibliotekę dokumentów
Global Trade Item Number
SKUGTIN
Z544825-1EA4061832116938