Z147915
Aldrich® solvent-still-head apparatus
head capacity 1,000 mL
About This Item
Kod UNSPSC:
41104812
NACRES:
NB.43
Polecane produkty
pojemność kolby
5,000 mL
pojemność nasadki
1,000 mL
Opis ogólny
24/40 joints. Includes still head, column with Raschig rings, Friedrichs condenser, one-neck round-bottom flask, stopper, and nitrogen-inlet adapter. Designed by Paul F. Hudrlik. May be used for continuous distillation.
Informacje prawne
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z147915-1EA | 4061832114668 |