Z247766
Aldrich® micro short-path distillation apparatus
Jacketed head
About This Item
Kod UNSPSC:
41104812
NACRES:
NB.43
Polecane produkty
Powiązane kategorie
Opis ogólny
Threaded 14/20 joints. Thermometers are included.
Replacement parts:
Z174521 - Standard head
Z247774 - Jacketed head
Z174548 - Cow receiver
Z174556 - Recovery flask, 5 mL
Z174564 - Distillation flask, 10 mL
Z174572 - Gas-inlet tube
Z561487 - Thermometer, 25 mm immr. L, -10 to 250 °C
Replacement parts:
Z174521 - Standard head
Z247774 - Jacketed head
Z174548 - Cow receiver
Z174556 - Recovery flask, 5 mL
Z174564 - Distillation flask, 10 mL
Z174572 - Gas-inlet tube
Z561487 - Thermometer, 25 mm immr. L, -10 to 250 °C
Powiązanie
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Informacje prawne
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
część zamienna
Numer produktu
Opis
Cennik
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Klienci oglądali również te produkty
Global Trade Item Number
SKU | GTIN |
---|---|
Z247766-1EA | 4061832131917 |