Z158380
Ace jacketed short-path distillation apparatus
Still body, plain
About This Item
Kod UNSPSC:
41104812
NACRES:
NB.43
Polecane produkty
producent / nazwa handlowa
Ace Glass 931750
Powiązane kategorie
Opis ogólny
For small quantities. Jacketed still body, with 10/18 thermometer joint, one-piece 14/20 cow receiver, and 10 mL distillation flask with 14/20 top opening. Order thermometer adapter Z125350 and thermometer separately.
Replacement parts:
Z158364, Jacketed still body, plain
Z158372, Jacketed still body, with Vigreux indents
Replacement parts:
Z158364, Jacketed still body, plain
Z158372, Jacketed still body, with Vigreux indents
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Klienci oglądali również te produkty
Global Trade Item Number
SKU | GTIN |
---|---|
Z158380-1EA | 4061832347974 |