Z223050
Extender for 500 and 750W ultrasonic processor
for horn 1/2 in.
About This Item
Kod UNSPSC:
41101500
NACRES:
NB.74
Polecane produkty
producent / nazwa handlowa
Sonics & Materials 630-0410
Opis ogólny
Screw into the threaded end of horn, extending the horn length by 5 in., for processing samples up to 7 in. down into narrow vessels. Often used with solvents or low surface tension liquids such as methylene chloride.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)