Kluczowe dokumenty
TEM-CF200F1-AU-UL
Sigma-Aldrich
Ultral-Thin Carbon Film Supported Gold Gilder Finder F1 Grids
size 200 mesh, box of 50
About This Item
Kod UNSPSC:
41111761
NACRES:
MA.24
rozmiar:
200 mesh
opakowanie:
box of 50
opakowanie
box of 50
rozmiar
200 mesh
Opis ogólny
A 3-4 nm ultral-thin film of pure carbon supported glider finder (F1) grids. Gold grid, 200 mesh. Gilder finder grids are locator grids with reference/index on the grid: 3.05mm diameter, 0.7 mil (18μm) thickness. F1 type of design has thick bars dividing regions into 6 thin bar areas, which are identified by a numeric system.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)