主要文書
ZUFPREUN8
Milli-Q
Installation kit for UF Pretreatment
Ultrafiltration pretreatment accessory
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41104214
NACRES:
JA.13
包装
pkg of 1 unit
適合性
for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX
詳細
This lab water purification accessory is used when interfacing the ultrafiltration (UF) pretreatment unit with Milli-Q® CLX/HX/HX-SD/HR 7000 systems.
アプリケーション
To be used when interfacing the UF pretreatment unit with a Milli-Q® CLX/HX/HX-SD/HR 7000 systems.
法的情報
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany