主要文書
ZLXL00P20
Milli-Q
ループ-20 mチューブ10x16、インター付き
For addition of single remote Point Of Delivery (POD) on the water distribution loop
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41104214
NACRES:
JA.13
包装
pkg of 1 unit
L
20 m , Tube
適合性
for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX
詳細
この20 mループオプションは、送水ループに1つのリモート採水ポイント(POD)を提供するために使用されます。
このキットには、20メートルの10x16 mmチューブに加えて、1つの採水ポイントを作るために必要な部品が含まれています。採水ポイントは、1台のアナライザーまたは他のデバイスに給水するために使用できます。
法的情報
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany