Documenti fondamentali
Z281387
Sigma-Aldrich
Basic laboratory brush kit
contains a variety of high quality laboratory brushes
About This Item
Codice UNSPSC:
47131605
NACRES:
NB.35
Descrizione generale
Convenience pack contains a variety of high quality laboratory brushes: 2 Micro tube, 2 Narrow tube, 2 Test tube, 1 Flask, 2 Cylinder, 1 Burette
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 6
1 of 6
Laboratory brushes Cylinder, L 11 in., brush diam. × L 2 3/8 in. × 5 in.
Z281271
Laboratory brushes
Laboratory brushes Burette, L 36 in., brush diam. × L 3/4 in. × 3 1/4 in.
Z281379
Laboratory brushes
Laboratory brushes Burette, L 30 in., brush diam. × L 1/2 in. × 3 1/4 in.
Z281360
Laboratory brushes
Laboratory brushes Pipette, L 17 in., brush diam. × L 3/16- 3/4 in. × 7 1/2 in.
Z281328
Laboratory brushes
Laboratory brushes Centrifuge, L 12 in., brush diam. × L 1 1/2 in. × 4 1/2 in.
Z281352
Laboratory brushes
Laboratory brushes Funnel, L 13 in., brush diam. × L 2 1/4 in. × 6 in.
Z281336
Laboratory brushes
Global Trade Item Number
SKUGTIN
Z281387-1EA4061833578735