Documenti fondamentali
Z281328
Laboratory brushes
Pipette, L 17 in., brush diam. × L 3/16- 3/4 in. × 7 1/2 in.
About This Item
Codice UNSPSC:
47131605
NACRES:
NB.35
Confezionamento
pkg of 4 ea
Lunghezza
17 in.
brush diam. × L
3/16- 3/4 in. × 7 1/2 in.
Descrizione generale
Brush materials resistant to acids, aromatic and aliphatic hydrocarbons, ketones, ethyl acetate esters, trichloroethylene, and hot water. FDA and USDA approved materials.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 6
1 of 6
Laboratory brushes Centrifuge, L 12 in., brush diam. × L 1 1/2 in. × 4 1/2 in.
Z281352
Laboratory brushes
Laboratory brushes Cylinder, L 11 in., brush diam. × L 2 3/8 in. × 5 in.
Z281271
Laboratory brushes
Laboratory brushes Test tube, L 9 in., brush diam. × L 3/4 in. × 3 1/4 in.
Z281255
Laboratory brushes
Laboratory brushes Test tube, L 10 in., brush diam. × L 1 3/8 in. × 4 in.
Z281263
Laboratory brushes
Global Trade Item Number
SKUGTIN
Z281328-1PAK4061826668511