Documenti fondamentali
BAF170680000
Cleanware swivel head bottle/flask brush
pack of 1 ea
Sinonimo/i:
bottle brush
About This Item
Codice UNSPSC:
41103200
NACRES:
NB.35
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F17068-0000
Descrizione generale
Use this brush to clean any type of deep vessel. Good for restaurant kitchens, laboratories with round-bottom flasks, repair shops, any facility needing dexterity and small size in the cleaning head.
Diameter of wooden head: 1.7025 in. x .8915 in. and the bristles are 1.578 in. x 0.74 in.
  • Head of brush swivels, reaching intricate curves and corners
  • Natural bristles set in wood will not scrape cleaned surface
  • Head fits into any container with a neck diameter of 24 mm (1 in.) or more.
  • Reach into deep vessels with 33 cm (13 in.) hardwood handle
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Lot/Batch Number
Ci dispiace, ma al momento non ci sono COA disponibili online per questo prodotto.
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 6
1 of 6
Laboratory brushes Test tube, L 10 in., brush diam. × L 1 3/8 in. × 4 in.
Z281263
Laboratory brushes
Laboratory brushes Narrow tube, L 12 in., brush diam. × L 1/2 in. × 3 in.
Z281301
Laboratory brushes
Laboratory brushes Funnel, L 13 in., brush diam. × L 2 1/4 in. × 6 in.
Z281336
Laboratory brushes
Laboratory brushes Burette, L 36 in., brush diam. × L 3/4 in. × 3 1/4 in.
Z281379
Laboratory brushes
Laboratory brushes Centrifuge, L 12 in., brush diam. × L 1 1/2 in. × 4 1/2 in.
Z281352
Laboratory brushes
Laboratory brushes Burette, L 30 in., brush diam. × L 1/2 in. × 3 1/4 in.
Z281360
Laboratory brushes