Documentos Principais
BR112810
BRAND® forcep with pointed ends
PMP, L 145 mm
Sinônimo(s):
Forceps
About This Item
Código UNSPSC:
41122405
NACRES:
NB.35
Materiais
PMP
Características
autoclavable at 121 C
fabricante/nome comercial
BRAND 112810
Comprimento
145 mm
Descrição geral
The BRAND® Forceps with Pointed Ends are precision tools designed for handling delicate materials in laboratory settings. Made from high-quality PMP (polymethylpentene), these forceps are lightweight yet durable, making them ideal for various applications. Designed to withstand autoclaving at 121 °C, ensuring effective sterilization for safe use in sterile environments.
Informações legais
BRAND is a registered trademark of BRAND GMBH + CO KG
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 5
1 of 5
Dressing tissue forceps L 6 in.
F4267
Dressing tissue forceps
Tweezers Style #7, sharp, hooked, PTFE-coated, anti-magnetic stainless steel
T6165
Tweezers
Stainless steel forceps Fine tip, straight
Z168777
Stainless steel forceps