Kluczowe dokumenty
Z381462
Heating blocks for Techne® Dri-Block® heater
holds 12, 15 mm tubes
About This Item
Kod UNSPSC:
41102404
NACRES:
NB.76
Właściwości
Holds 12 x 15 mm tubes
producent / nazwa handlowa
Techne F3506
Opis ogólny
Interchangeable blocks machined of aluminum for rapid heat transfer. Close tolerances assure efficient, rapid heating. Each block has a threaded thermometer hole for independent temperature monitoring, which also fits the block extraction tool.
Informacje prawne
Dri-Block is a registered trademark of Techne, Inc.
Techne is a registered trademark of Techne, Inc.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)