Kluczowe dokumenty
Z219355
Stainless steel 304 syringe needle
gauge 19, L 10 in.
About This Item
Kod UNSPSC:
41122002
eCl@ss:
32070500
NACRES:
NB.03
Polecane produkty
opis
Standard Hub with 304 Stainless Steel Tubing
opakowanie
pack of 1 ea
producent / nazwa handlowa
Cadence Science
Długość
10 in.
grubość
19
Powiązane kategorie
Opis ogólny
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Standard Hub with 304 Stainless Steel Tubing
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z219355-1EA | 4061832530741 |