Z122904
Ampule breaker/collar
For 1-4 mL ampules
About This Item
Kod UNSPSC:
41121807
NACRES:
NB.43
Polecane produkty
Opis ogólny
How to Use:
1. Slip polyethylene breaker/ collar over top of ampule.
2. Grip bottom of ampule with one hand and breaker/collar with other.
3. Gently snap top of ampule off.
4. Discard collar and ampule top.
1. Slip polyethylene breaker/ collar over top of ampule.
2. Grip bottom of ampule with one hand and breaker/collar with other.
3. Gently snap top of ampule off.
4. Discard collar and ampule top.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Klienci oglądali również te produkty
Global Trade Item Number
SKU | GTIN |
---|---|
Z122904-1000EA | |
Z122904-100EA | 4061833307137 |
Z122904-10X100EA |