Kluczowe dokumenty
MT30014099
Mettler Toledo InLab® trace kit
About This Item
Kod UNSPSC:
41115603
NACRES:
NB.79
Materiały
titanium shaft
opis
Mini-LTW connector
producent / nazwa handlowa
Mettler-Toledo 30014099
zakres pomiaru przewodności
0.0001-1000 μS/cm
dł. × szerokość
170 mm × 12 mm
rozmiar kabla
1.8 m
Opis ogólny
Mettler Toledo InLab® Trace Kit includes both probe (30014097) and Flow Cell (30014098).
Low conductivity measurement requires special attention. When measuring pure or ultra-pure water there are several sources of potential error such as carbon dioxide from air and temperature inaccuracy: The digital conductivity sensor Mettler Toledo InLab® Trace has a high precision temperature probe. The conductivity and temperature signals are digitalized and combined through a special algorithm for unmatched conductivity accuracy.
The optional flow-cell made of polypropylene provides the perfect protection from carbon dioxide. Because it does not contain a single piece of conducting material, any interference is excluded.
Using the Flow Cell the InLab® Trace the highest repeatability and reproducibility ever can be achieved.
The Flow Cell InLab® Trace was built at an angle to prevent air bubbles being trapped. Your workflow is optimized to the max. Enjoy the peace of mind.
For pure water we also offer a special temperature compensation mode to increase the reliability of your readings.
Each sensor is supplied with a certificate specifying the actual cell constant which makes calibration superfluous.
  • High precision, digitalized temperature and conductivity readings
  • Certified cell constant which makes calibration superfluous
Informacje prawne
InLab is a registered trademark of Mettler-Toledo, Inc.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)