Merck
GCによるポテトチップの分析、SPB®-1 SULFUR、SPME処理、 50/30 μm DVB/Carboxen/PDMS使用
GCによるポテトチップの分析、SPB®-1 SULFUR、SPME処理、 50/30 μm DVB/Carboxen/PDMS使用 suitable for GC, application for SPME
材料
SPMEファイバー
製品番号
詳細
価格
SPMEファイバーアセンブリー ジビニルベンゼン/カルボキセン/ポリジメチルシロキサン (DVB/CAR/PDMS)
50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Standard
製品番号
詳細
価格
デカナール
≥98% (GC), liquid
酪酸メチル
analytical standard
ヘキサン酸
analytical standard
オクタン酸
analytical standard
オクタン
analytical standard
ヘプタナール
≥95%, FCC, FG
ノナナール
analytical standard
ヘキサナール
analytical standard
Butyl hexanoate
analytical standard
オクタナール
analytical standard
ヘプタン酸
analytical standard
分析カラム
製品番号
詳細
価格
SPB®-1 SULFURキャピラリーGCカラム
L × I.D. 30 m × 0.32 mm, df 4.00 μm
関連製品
製品番号
詳細
価格
SPMEファイバーホルダー
for use with Manual Sampling
SPMEファイバーホルダー
for use with CTC CombiPAL, Gerstel MPS2
CONDITIONS
sample/matrix
3 g crushed potato chips in 15 mL vial
SPME fiber
DVB/Carboxen/PDMS, 50/30 μm, StableFlex (57328-U)
extraction
headspace, 65 °C, 20 min, with stirring
desorption process
3 min, 250 °C
column
SPB-1 SULFUR, 30 m × 0.32 mm I.D., 4.0 μm (24158)
inj. temp.
250 °C
oven
45 °C (1.5 min) to 250 °C at 12 °C/min, hold 10 min
detector
quadrupole mass spectrometer, m/z = 35-290 at 0.6 sec/scan
carrier gas
helium, 40 cm/sec
injection
Splitless/split, closed 2 min
liner
0.75 mm liner
sample
3g crushed potato chips in 15 mL vial
詳細
アナリシスノート
Potato chips can contain up to 50% oil and are subject to rancidity. Aldehydes, particularly hexanal and heptanal, are formed from fatty acids that are cleaved at the double bonds when exposed to ultraviolet light and air. A package of 3-year-old potato chips was analyzed using SPME and compared to a fresh package. The presence of aldehydes indicated that the older chips were rancid. However, due to the age of the chips, further oxidation of the aldehydes occurred that resulted in the formation of hexanoic and heptanoic acids. A Carboxen/PDMS/DVB fiber was inserted into the headspace of the potato chip samples at 65 °C for 20 minutes with agitation. The fiber was then analyzed using GC/MS on a SPB-1 SULFUR column. This non-polar column has a thick film and is ideal for smaller molecules.
法的情報
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany