GCによる 1-メトキシ-2-プロピルアミンエナンチオマー(N-クロロアセチル誘導体)の分析、Astec® CHIRALDEX™ B-PH
![GCによる 1-メトキシ-2-プロピルアミンエナンチオマー(N-クロロアセチル誘導体)の分析、Astec® CHIRALDEX™ B-PH suitable for GC](/deepweb/assets/sigmaaldrich/product/images/308/915/c1f4ad16-2257-4eb6-bbc1-d42df77ea32f/800/c1f4ad16-2257-4eb6-bbc1-d42df77ea32f.jpg)
CONDITIONS
column
Astec CHIRALDEX B-PH, 30 m x 0.25 mm I.D., 0.12 μm (71023AST)
oven
120 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 24 psi
injection
1 μL, 80:1 split
sample
3 mg/mL in methanol
peaks 1 & 2: 1-methoxy-2-propylamine enantiomers (N-chloroacetyl derivatives)