ZSTWINLX2
Tank Level Display Accessory
Reports the PE Tank level signal feeding a Milli-Q® IQ/EQ 7000 and Advantage/Reference systems
About This Item
Código UNSPSC:
41104200
NACRES:
JA.13
Produtos recomendados
entrada CA/CC
28 V
embalagem
pkg of 1 unit
compatibilidade
for use with Milli-Q® EQ 7000
for use with Milli-Q® Advantage A10
for use with Milli-Q® IQ 7000
for use with Milli-Q® Reference
Descrição geral
This water system accessory transfers PE tank level information to the Milli-Q® Advantage or Milli-Q® Reference and Milli-Q® IQ/EQ 7000 water purification systems.
Aplicação
Tank level display accessory - Duplicates tank level signal.
Informações legais
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany