Documentos Principais
CDRCHKIT2
Milli-Q
Cartucho de osmose reversa
High fouling RO Kit for RiOs 50 and Elix® 35 Water Purification System
About This Item
Código UNSPSC:
41104214
NACRES:
JA.13
embalagem
pkg of 1 unit
compatibilidade
for use with Elix®
for use with RiOs
Descrição geral
The Reverse Osmosis membrane removes 95%-99% of contaminants including ions, particulates, bacteria, and organic molecules. For use with, Elix® 35 and RiOs 50 water purification systems.
Aplicação
The RO water delivered from Elix® and RiOs systems is used in laboratories, mainly for applications such as general rinsing, feedwater to washing machines and autoclaves, or as a feed to pure and ultrapure water purification systems.
Informações legais
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany