About This Item
Código UNSPSC:
40141600
NACRES:
NB.51
Produtos recomendados
faixa de medição de pressão
≤3,000 psig
Tamanho interno da CGA
180
Descrição geral
- Use with SS lecture-bottle regulators when regulating corrosive gases
- 316 SS with CGA inlet/outlet connections
- Diaphragm-seal purge valve with 1/4 in. NPTM inlet
- Check-valve at purge-inlet prevents backflow of gas
Valve installs between lecture-bottle and regulator to purge system of air, moisture, and process gas. Minimizes gas hydrolysis and corrosion damage to equipment.
Purge system with dry inert gas before introducing corrosive gases and again immediately after use. PTFE sealing washers are NOT included.
Note: Order spare washers, listed separately.
Outras notas
See Technical Information Bulletin AL-151 Gas Regulators: Selection, Installation and Operation.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
Z151661-1EA | 4061833639832 |