Z562483
Aldrich® corrosive gas regulator with electroless nickel-plated body and cross purge assembly
inlet CGA 180
About This Item
Código UNSPSC:
41112404
NACRES:
NB.51
Produtos recomendados
Materiais
316L stainless steel diaphragm (PTFE-lined)
Monel® nozzle
descrição
Captured vent
PTCFE seat and Viton® seals
delivery pressure measuring range
0-80 psig
faixa de medição da pressão de entrada
3000 psig
entrada da CGA
180
NPTM de saída
1/4 in.
Descrição geral
The combination of Monel® regulator components and proper purging with the cross-purge assembly makes these units virtually impervious to attack from strong acid forming gases that include hydrogen chloride, chlorine, hydrogen fluoride, hydrogen bromide, and silicon tetrafluoride.
Cross-purge assembly eliminates the escape of toxic and flammable gases when changing cylinders and permits purging system with dry, inert gas to prevent corrosion due to gas hydrolysis.
Specifications:
Request Technical Information Bulletin No. AL-202 for cross-purge techniques.
Cross-purge assembly eliminates the escape of toxic and flammable gases when changing cylinders and permits purging system with dry, inert gas to prevent corrosion due to gas hydrolysis.
Specifications:
- Single-stage
- Electroless nickel-plated body
- Stainless steel diaphragm
- 316L Stainless steel cross-purge with color-coded 1/4 turn valves with 1/4 in. NPTM outlets
Request Technical Information Bulletin No. AL-202 for cross-purge techniques.
Outras notas
See Technical Information Bulletin AL-151 Gas Regulators: Selection, Installation and Operation.
Informações legais
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Monel is a registered trademark of Huntington Alloys Corp.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
Z562483-1EA | 4061837636783 |