CAD6030
1-way stopcock
male Luer lock to 8 mm hose end
About This Item
Código UNSPSC:
41122000
NACRES:
NB.03
Produtos recomendados
embalagem
pack of 1 ea
fabricante/nome comercial
Cadence Science 6030
Categorias relacionadas
Descrição geral
Stopcocks are precision-machined, individually lapped, and carefully mated to maintain pressure characteristics. These stopcocks are easily integrated with other stopcocks, manifolds, adapters, tubing, needles, and glass syringes.
- Made from nickel plated brass or stainless steel
- Tested to 45 psi water pressure
- High pressure stopcocks are rated to 290 psi
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
CAD6030-1EA | 4061832517285 |