Z569925
Chem-FLEX™ transfer line with male and female Luer connectors
tube L 30 in.
About This Item
Código UNSPSC:
41121815
NACRES:
NB.35
Produtos recomendados
tubo C
30 in.
Descrição geral
One male and one female Luer lock connector with chrome-plated brass bodies are connected to Chem-Flex 106 tubing with nylon clamps. Chem-Flex tubing was developed by Aldrich for the safe transfer of aggressive solvents, air-sensitive liquids, and gases.
Chem-Flex Tubing Features:
Chemically inert, thin-wall PTFE inner tube bonded to a clear PVC outer tube for extra strong, flexible transfer lines that are resistant to crushing or kinking.
Applications:
Chem-Flex Tubing Features:
Chemically inert, thin-wall PTFE inner tube bonded to a clear PVC outer tube for extra strong, flexible transfer lines that are resistant to crushing or kinking.
- Luer fittings on ends of tubing allow easy connection to
- Stopcocks and valves
- Needles of all lengths and gauges
- Syringes and pumps
Applications:
- Pressure transfer of liquids- Using Luer needles, stopcocks
- Reagent addition to reactions- Using Luer needle, stopcock, syringe or syringe pump
Informações legais
Chem-Flex is a trademark of Sigma-Aldrich Co. LLC
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
Z569925-1EA | 4061826709627 |