SYND161420
Synthware™ cow style distillation receiver
joint: ST/NS 14/20
Synonym(e):
Synthware D161420, cow type distilling receiver
Alle Fotos(1)
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.51
Empfohlene Produkte
Allgemeine Beschreibung
Synthware™ distillation cow receiver is designed for microscale applications. It features a standard taper joint (14/20) and four drip-tip joints of the same size. The receiver ensures collection of the condensate in four flasks progressively.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)