Merck
SYND151814
Synthware short path distillation head
joint: ST/NS 14/20
Synonym(e):
Synthware D151814, Short path head
About This Item
UNSPSC-Code:
41104812
NACRES:
NB.51
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware D151814
Allgemeine Beschreibung
Short-path design for microscale application, used with 25 mm immersion thermometer, and D27 distillation receiver.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)