Merck
SYND251019
Synthware Vigreux microscale distillation head
joint: ST/NS 19/22, immersion depth 25 mm
Synonym(e):
Synthware D251019, short path, distilling head
About This Item
UNSPSC-Code:
41104812
NACRES:
NB.51
Verbindung
joint: ST/NS19/22
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware D251019
Eintauchtiefe
25 mm
Allgemeine Beschreibung
With Vigreux Indentations, used with 25 mm immersion thermometer, and D27 distillation receiver.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)