Merck
SYND271420
Synthware distillation distribution adapter
joint: ST/NS 14/20
Synonym(e):
Synthware D271420, short path distillation receiver
About This Item
UNSPSC-Code:
41104812
NACRES:
NB.51
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware D271420
Verwandte Kategorien
Allgemeine Beschreibung
With the outer and inner joint at 135° angle, used with short-path.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)