Z738719
Glow Writer™ autoradiography pen
replacement nibs
About This Item
Kod UNSPSC:
41105341
NACRES:
NB.31
Polecane produkty
producent / nazwa handlowa
Diversified Biotech GNIB-1000
Opis ogólny
Glow Writer is a phosphorescent marking pen for annotating or marking chemiluminescent and isotope-labeled substrates for X-ray film autography. The unique marker delivers a phosphor-containing marking ink through a fine point porous nib. Before exposure to X-ray film, the phosphorescent writing is excited by bright room light. A phosphor ink having an optimum concentration of phosphor and a relatively short afterglow half-life (approx. 2-3 minutes) has been selected so that upon exposure to X-ray film, autographic exposures of almost any duration will be easily readable on the film. The fine point nib is easily replaceable if it dries out.
Informacje prawne
Glow Writer is a trademark of Diversified Biotech
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)