Kluczowe dokumenty
AXYPDW20CSIND
Deep well plate
individually wrapped, size 96 wells, size 2.0 mL, pkg of 30 ea, sterile
Synonim(y):
Corning cell culture plate
About This Item
Kod UNSPSC:
41121800
NACRES:
NB.22
Materiały:
round bottom wells
rozmiar:
2.0 mL
96 wells
Materiały
round bottom wells
sterylność
sterile
opakowanie
pkg of 30 ea
producent / nazwa handlowa
Axygen Inc P-DW-20-C-S-IND
Corning
rozmiar
2.0 mL
96 wells
Opis ogólny
  • Axygen® deep well assay plates are ideal for use in applications including sample collection, in vitro growth chambers and long-term storage.
  • Chemically resistant and withstands temperatures down to -80°C.
  • Ultra-low profile design for reduced space requirements.
  • Flat surface for efficient sealing using heat sealing film.
Informacje prawne
Axygen is a registered trademark of Corning, Inc.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)