Kluczowe dokumenty
Z531847
Sigma-Aldrich
Tubing adapter with Torion connection and stopcock
PTFE stopcock, O.D. 3/16 in., male joint: ST/NS 19/22
About This Item
Kod UNSPSC:
41121815
NACRES:
NB.35
Materiały
PTFE stopcock
Połączenie
male joint: ST/NS19/22
śr. zewn.
3/16 in.
4.8 mm
Opis ogólny
PTFE stopcocks are threaded for additional safety and have a 2 mm bore for 3/16 in. tubing connection and 4 mm bore for 5/16 in. and 1/2 in. tubing connections. 5/16in. O.D fits a #8 Torion, and 1/2in. O.D. fits a #14 Torion.
Informacje prawne
Torion is a trademark of Bibby Sterlin Ltd.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)