Z275336
Cannon-Fenske viscometer tube
size 200
About This Item
Kod UNSPSC:
41111700
NACRES:
PB.36
Polecane produkty
rozmiar
200
opis
constant ~ 0.1 cSt/s
producent / nazwa handlowa
Ace Glass 798843
zakres pomiaru lepkości
20-100 cSt
Powiązane kategorie
Opis ogólny
Obtain kinematic viscosities using tubes designed for ASTM Method D445 and D2515. Viscosities can be measured with a precision of ±0.2% quickly and easily.
Requires approximately 7 mL of sample. Certificate of Calibration supplied.
Requires approximately 7 mL of sample. Certificate of Calibration supplied.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z275336-1EA | 4061832349213 |