Z275271
Cannon-Fenske viscometer tube
size 50
About This Item
Kod UNSPSC:
41111700
NACRES:
PB.36
Polecane produkty
rozmiar
50
opis
constant ~ 0.004 cSt/s
producent / nazwa handlowa
Ace Glass 798835
zakres pomiaru lepkości
0.8-4 cSt
Powiązane kategorie
Opis ogólny
Obtain kinematic viscosities using tubes designed for ASTM Method D445 and D2515. Viscosities can be measured with a precision of ±0.2% quickly and easily.
Requires approximately 7 mL of sample. Certificate of Calibration supplied.
Requires approximately 7 mL of sample. Certificate of Calibration supplied.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z275271-1EA | 4061832349176 |