Z261718
Luer-to-Closed end stopper
Micro-Mate® female Luer, 316 stainless steel
About This Item
Kod UNSPSC:
41122500
NACRES:
NB.03
Polecane produkty
Materiały
316 stainless steel
producent / nazwa handlowa
Popper 6450
Powiązane kategorie
Opis ogólny
Hardware specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes.
316 SS fittings are noncontaminating, easy to clean and resistant to corrosion. Chrome-plated brass connectors offer good performance at an economical price. Water tested at 45 psi.
316 SS fittings are noncontaminating, easy to clean and resistant to corrosion. Chrome-plated brass connectors offer good performance at an economical price. Water tested at 45 psi.
Informacje prawne
Micro-Mate is a registered trademark of Cadence Science
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Klienci oglądali również te produkty
Global Trade Item Number
SKU | GTIN |
---|---|
Z261718-1EA | 4061826668320 |