Kluczowe dokumenty
Z261270
Stainless steel 316 syringe needle, noncoring point
gauge 20, L 1 in.
About This Item
Kod UNSPSC:
42142600
NACRES:
NB.03
Polecane produkty
opis
Micro-Mate Hub with 316 Stainless Steel Tubing
opakowanie
pack of 1 ea
producent / nazwa handlowa
Popper 9862
Długość
1 in.
grubość
20
Opis ogólny
Specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. Female Luer hubs and needles are made of 316 SS. Noncontaminating, easy to clean and resistant to corrosion.
Micro-Mate Hub with 316 Stainless Steel Tubing
Micro-Mate Hub with 316 Stainless Steel Tubing
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)