Merck
Z261262
Stainless steel 316 syringe needle, noncoring point
gauge 22, L 6 in.
About This Item
Kod UNSPSC:
42142600
NACRES:
NB.03
opis
Micro-Mate Hub with 316 Stainless Steel Tubing
opakowanie
pack of 1 ea
producent / nazwa handlowa
Popper 9866
Długość
6 in.
grubość
22
Opis ogólny
Specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. Female Luer hubs and needles are made of 316 SS. Noncontaminating, easy to clean and resistant to corrosion.
Micro-Mate Hub with 316 Stainless Steel Tubing
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Odwiedź Bibliotekę dokumentów
Global Trade Item Number
SKUGTIN
Z261262-1EA4061832530772