Kluczowe dokumenty
Z222844
Quick-connect hose coupling
for tubing i.d., 1/4 in., Coupling
About This Item
Kod UNSPSC:
41121815
NACRES:
NB.35
Właściwości
for tubing i.d. 1/4 in.
Opis ogólny
Nickel-plated brass coupling and nipple. Simple push-to-connect operation. Suitable for both pressure and vacuum applications. Max. pressure: 508 psi, min. burst pressure: 2,030 psi. Temp. range: -40 to 100 °C (-40 to 212 °F).
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)