Kluczowe dokumenty
Z154245
Compressor-type tubing clamp
heavy-duty, nickel-plated brass
About This Item
Kod UNSPSC:
41105108
NACRES:
NB.35
otwór szer. × wys.
35 mm × 30 mm
Opis ogólny
  • Heavy-duty, nickel-plated brass
  • Opening: 35 W × 30 H (mm)
  • Lower bar held in place by threaded cap; may be swung out to insert tubing
  • 6 mm screw with 35 mm knurled finger grip
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)