Kluczowe dokumenty
Z113050
Stainless steel 304 syringe needle
gauge 17, L 3 1/2 in.
About This Item
Kod UNSPSC:
41122002
NACRES:
NB.03
opis
Standard Hub with 304 Stainless Steel Tubing
opakowanie
pack of 1 ea
producent / nazwa handlowa
Cadence Science
Długość
3 1/2 in.
grubość
17
Opis ogólny
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)