Kluczowe dokumenty
CAD9930
Animal feeding needle, sterile, disposable, PTFE
size 14G, L × diam. 4.7 in. × 2.9 mm , ball
About This Item
Kod UNSPSC:
42142609
NACRES:
NB.03
rozmiar
14G
opakowanie
pkg of 20 x 5 ea (5 needles per sterile Tyvek peel pouch - 20 pouches per box)
dł. × śr.
4.7 in. × 2.9 mm , ball
Opis ogólny
  • PTFE tube is softer than other tubing types, very flexible, and translucent for easy confirmation of reagent delivery
  • Plastic Luer hubs and soft silicone tips
  • Non-toxic and no-pyrogenic
  • Packaged 5 needles per sterile Tyvek peel pouch - 20 pouches per box
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)