Kluczowe dokumenty
BR705932
BRAND® Transferpette® S Multichannel Pipette
capacity (30-300 μL), 12-channel, adjustable volume, M12-300, DE-M
Synonim(y):
Pipety, Pipety tłokowe, Transferpettor, Zasada wyporu dodatniego, Zestaw naprawczy
About This Item
Kod UNSPSC:
41121500
NACRES:
NB.01
Właściwości
12-channel adjustable volume
opakowanie
case of 1 ea
producent / nazwa handlowa
BRAND 705932
pojemność
(30-300 μL)
Opis ogólny
The Transferpette S air displacement pipette allows you to work efficiently and ergonomically with both small and large volumes. Low operating forces and a short stroke ensure relaxed pipetting, even during long pipetting series. Due to its perfect ergonomics, the Transferpette S provides a comfortable grip in any hand position, whether you are right- or left-handed, or have big or small hands. Changing your grip is not necessary, since the microliter pipette provides true one-hand operation: set the volume, pipette, eject the tip.The volume-change protection is also easy to operate with gloves on and protects your set volume against accidental adjustment. The Transferpette S also features Easy Calibration technology, which makes calibration a breeze. Calibration can be carried out without tools. A compatibility table shows which tips fit which device, so that you always order the correct pipette tips.
Overview- Volume range of single-channel pipette: from 0.1 μl to 10 ml- Volume range of multi-channel pipette: from 0.5 μl to 300 μl- Short stroke and low operating forces- Universal tip cone- Completely autoclaving at 121 μC- One-hand operation for right- and left-handed users- Color code on volume adjustment wheel, shaft and volume-change protection- Easy insertion and ejection of tips due to O-rings on multi-channel pipettes- Can be calibrated easily and without tools thanks to Easy Calibration technology- Slim shaft for sampling from narrow vessels (centrifuge tubes)- Operating temperature from +15 μC to +40 μC- Vapor pressure up to 500 mbar- Viscosity up to 260 mPa s
Efficient: One-hand operation
Flexible: ergonomic design for all hand sizes
Reliable: volume-change protection prevents accidental adjustments
Cechy i korzyści
  • Wydajność: obsługa jedną ręką
  • Elastyczność: ergonomiczna konstrukcja dla wszystkich rozmiarów dłoni
  • Niezawodność: zabezpieczenie przed przypadkową zmianą głośności
Informacje prawne
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Odwiedź Bibliotekę dokumentów
Global Trade Item Number
SKUGTIN
BR705932-1EA4061841857372