Merck
HPLCによる5,5-ジメチル-4-フェニル-2-オキサゾリジノンエナンチオマーの分析、CHIROBIOTIC® T
HPLCによる5,5-ジメチル-4-フェニル-2-オキサゾリジノンエナンチオマーの分析、CHIROBIOTIC® T application for HPLC
材料
分析カラム
製品番号
詳細
価格
(S)-(+)-5,5-Dimethyl-4-phenyl-2-oxazolidinone
98%
Astec® CHIROBIOTIC® T Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] 0.5% triethylamine acetate, pH 4.1; [B] methanol; (80:20, A:B)
flow rate
1 mL/min
column temp.
ambient
detector
UV, 254 nm
injection
5 μL
sample
5 mg/mL in methanol