GCによるブピバカインエナンチオマーの分析、Astec® CHIRALDEX™B-PH
CONDITIONS
column
Astec CHIRALDEX B-PH, 10 m x 0.25 mm I.D., 0.12 μm (custom)
gradient
130 °C, 2 °C/min to 170 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 12 psi
sample
peaks 1 & 2: bupivacaine enantiomers