GC Analysis of Volatile Flavor Compound Enantiomers on Astec® CHIRALDEX™ G-TA
CONDITIONS
column
Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 μm (73033AST)
oven
60 °C, 2 °C/ min to 160 °C (hold 20 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 cm/sec, set at 75 °C
injection
1 μL, split 100:1