特徴
to hold 20 x 5.0 mL Eppendorf tubes
aerosol tight
fixed angle rotor
with Eppendorf QuickLock® rotor lid
包装
pack of 1 ea
メーカー/製品名
Eppendorf® 5895130003 (Global)
パラメーター
25,062 × g max. RCF
テクニック
centrifugation: suitable
適合性
for use with centrifuge models 5920R
類似した製品をお探しですか? 訪問 製品比較ガイド
関連するカテゴリー
詳細
The large Centrifuge 5920 R is a benchtop centrifuge that combines exceptional capacity with a compact and ergonomic design. It has similar dimensions to refrigerated competitor models in the 3-liter segment, but has an outstanding capacity of up to 4 x 1,000 mL or 52 x 50 mL tubes. Optimize high-throughput centrifugation and make your daily work easier.
法的情報
Eppendorf is a registered trademark of Eppendorf AG
最新バージョンのいずれかを選択してください:
Global Trade Item Number
カタログ番号 | GTIN |
---|---|
EP5895130003-1EA | 4061835079605 |