Merck
EP0030089502
Eppendorf® ViscoTip, 10 mL, Eppendorf Quality
pack of 100 ea (4 bags of 25 tips)
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41121600
NACRES:
NB.02
最大容量:
10 mL
容量:
(10 mL)
無菌性:
non-sterile
包装:
pack of 100 ea (4 bags of 25 tips)
メーカー/製品名:
Eppendorf® 0030089502
物質
orange
無菌性
non-sterile
包装
pack of 100 ea (4 bags of 25 tips)
メーカー/製品名
Eppendorf® 0030089502
最大容量
10 mL
容量
(10 mL)
特徴および利点
  • Tips are specialized for handling highly viscous liquids that have a dynamic viscosity from 200 mPa*s to 14,000 mPa*s
  • Ideal for dispensing volumes ranging between 100 µL to 10 mL, with 10 µL increments
  • New tip design increases ergonomics, and working speed and reduces energy consumption
  • Save time on calculations with the Multipette® M4/E3/E3x, which automatically recognizes tips and calculates volume.
  • Free of leachable such as slip agents, biocides, and plasticizers that interfere with experimental results
  • ViscoTips come with a distinct color-coded double ring, and continuous volume scale, making it easier to differentiate them from Combitips advanced®
法的情報
Combitips advanced is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Quality is a trademark of Eppendorf AG
Multipette is a registered trademark of Eppendorf AG
最新バージョンのいずれかを選択してください:
試験成績書(COA)