コンテンツへスキップ
Merck
すべての画像(2)

主要文書

安全性情報

AB9878

Sigma-Aldrich

Anti-MAGI-2 Antibody

Chemicon®, from rabbit

別名:

Membrane Associated Guanylate Kinase, WW and PDZ domain containing 2, S-SCAM, Synaptic Scaffolding Molecule

ログイン組織・契約価格を表示する


About This Item

UNSPSCコード:
12352203
eCl@ss:
32160702
NACRES:
NA.41
クローン:
polyclonal
application:
ICC
IHC
IP
WB
化学種の反応性:
mouse
テクニック:
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
2

由来生物

rabbit

抗体製品の状態

purified antibody

抗体製品タイプ

primary antibodies

クローン

polyclonal

化学種の反応性

mouse

メーカー/製品名

Chemicon®

テクニック

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBIアクセッション番号

UniProtアクセッション番号

輸送温度

wet ice

ターゲットの翻訳後修飾

unmodified

遺伝子情報

human ... MAGI2(9863)

詳細

The spatial coordination of neurotransmitter receptors with other postsynaptic signaling and structural molecules is regulated by a diverse array of cell-specific scaffolding proteins. The synaptic trafficking of AMPA receptors by the stargazin protein in some neurons, for example, depends on specific interactions between the C terminus of stargazin and the PDZ [postsynaptic density-95 (PSD-95)/Discs large/zona occludens-1] domains of membrane-associated guanylate kinase scaffolding proteins PSD-93 or PSD-95. Stargazin [Cacng2 (Ca2+ channel gamma2 subunit)] is one of four closely related proteins recently categorized as transmembrane AMPA receptor regulating proteins (TARPs) that appear to share similar functions but exhibit distinct expression patterns in the CNS. MAGI-2 (membrane associated guanylate kinase, WW and PDZ domain containing 2) has been identified as a novel candidate interactor with the cytoplasmic C termini of the TARPs. MAGI-2 [also known as S-SCAM (synaptic scaffolding molecule)] is a multi-PDZ domain scaffolding protein that interacts with several different ligands in brain, including PTEN (phosphatase and tensin homolog), dasm1 (dendrite arborization and synapse maturation 1), dendrin, axin, beta- and delta-catenin, neuroligin, hyperpolarization-activated cation channels, beta1-adrenergic receptors, and NMDA receptors. MAGI-2 coimmunoprecipitates with stargazin in vivo. Expression of stargazin recruites MAGI-2 to cell membranes and cell-cell contact sites in transfected HEK-293T cells dependent on the presence of the stargazin -TTPV motif. MAGI-2 represents a strong candidate for linking TARP/AMPA receptor complexes to a wide range of other postsynaptic molecules and pathways and advance our knowledge of protein interactions at mammalian CNS synapses.

特異性

Cat. # AB9878 recognizes MAGI-2
Reactivity with other species has not been determined.

免疫原

GST-tagged MAGI-2

アプリケーション

This Anti-MAGI-2 Antibody is validated for use in IC, IH, IP, WB for the detection of MAGI-2.

品質

Routinely evaluated by Western Blotting on mouse brain tissue lysate.

Western Blotting Analysis: 1:500 dilution of this lot was used to detect MAGI-2 on mouse brain lysate.

ターゲットの説明

~140 kDa

物理的形状

Format: Purified
Purified antibody in 0.1M Tris-Glycine (pH7.4) 15mM NaCl with 0.05% NaN3.

保管および安定性

Stable for 1 year at 2-8°C from date of receipt.
Handling Recommendations: Prior to removing the cap, gently centrifuge the vial and gently mix the solution.

アナリシスノート

Control
Mouse brain

その他情報

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

法的情報

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

適切な製品が見つかりませんか。  

製品選択ツール.をお試しください

保管分類コード

12 - Non Combustible Liquids

WGK

WGK 1

引火点(°F)

Not applicable

引火点(℃)

Not applicable


適用法令

試験研究用途を考慮した関連法令を主に挙げております。化学物質以外については、一部の情報のみ提供しています。 製品を安全かつ合法的に使用することは、使用者の義務です。最新情報により修正される場合があります。WEBの反映には時間を要することがあるため、適宜SDSをご参照ください。

Jan Code

AB9878:


試験成績書(COA)

製品のロット番号・バッチ番号を入力して、試験成績書(COA) を検索できます。ロット番号・バッチ番号は、製品ラベルに「Lot」または「Batch」に続いて記載されています。

以前この製品を購入いただいたことがある場合

文書ライブラリで、最近購入した製品の文書を検索できます。

文書ライブラリにアクセスする

Naomi Tanga et al.
Glia, 67(5), 967-984 (2019-01-23)
Protein tyrosine phosphatase receptor type Z (PTPRZ) maintains oligodendrocyte precursor cells (OPCs) in an undifferentiated state. The inhibition of PTPase by its ligand pleiotrophin (PTN) promotes OPC differentiation; however, the substrate molecules of PTPRZ involved in the differentiation have not

ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.

製品に関するお問い合わせはこちら(テクニカルサービス)