詳細
molecule(s) 1
メーカー/製品名
Molymod
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
The mushroom-shaped hydrogen Atomlink™ components and the short (non-visible) bonding links provide compact, semi-space-filling models with an internuclear scale of 1.45 cm per Angstrom. Polymer models include long, flexible links for demonstration of the mechanism of polymerization.
法的情報
Atomlink is a trademark of Spiring Enterprises Ltd.
Molymod is a registered trademark of Spiring Enterprises Ltd.
最新バージョンのいずれかを選択してください:
Global Trade Item Number
カタログ番号 | GTIN |
---|---|
Z544728-1SET | 4061833078341 |