Merck
BAF182370000
Dialysis bag holder clip for beakers
pack of 6 ea
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41123100
NACRES:
NB.51
物質
polypropylene
特徴
autoclavable
包装
pack of 6 ea
メーカー/製品名
Bel-Art F18237-0000
詳細
The bag holder is resistant to chemicals and may be autoclaved.
  • Suspend dialysis/osmosis bag in fluid
  • Sturdy holder clips onto the side of an open container with a wall up to 1/4 in. thick
  • Holder has room for sample identification marks
最新バージョンのいずれかを選択してください:
試験成績書(COA)