Merck
TLC Analysis of Caffeine in Coffee Samples, Quantification of Analyte Concentration
TLC Analysis of Caffeine in Coffee Samples, Quantification of Analyte Concentration application for TLC
Materiali
Accessorio
N° Catalogo
Descrizione
Determinazione del prezzo
Bottle-type sprayer
capacity 240 mL
Rectangular TLC developing tanks, complete
L × H × W 27.0 cm × 26.5 cm × 7.0 cm
Sigma TLC spray box
Spectroline® E-Series UV lamp
output8 W, AC/DC input 115 V AC
Rectangular TLC developing tanks, complete
L × H × W 7.5 cm × 15.5 cm × 8.0 cm
Hamilton® TLC syringes
1702, volume 25 μL, needle size 22s ga, needle L 51 mm (2 in.),
coated tip
Spectroline® E-Series UV lamp
output6 W, AC/DC input 120 V AC
Hamilton® TLC syringes
1705, volume 50 μL, needle size 22 ga, needle L 51 mm (2 in.),
coated tip
Hamilton® TLC syringes
1701, volume 10 μL, needle size 26s ga, needle type, fixed, needle L 51 mm (2 in.),
coated tip
Rectangular TLC developing tanks, complete
L × H × W 17.5 cm × 16.0 cm × 8.2 cm
Spectroline® E-Series UV lamp
output6 W, AC/DC input 230 V AC
Spectroline® E-Series UV lamp
output4 W, AC/DC input 230 V AC
Spectroline® E-Series UV lamp
output8 W, AC/DC input 230 V AC
Latch-lid TLC developing chambers
For use with 10 cm × 10 cm plates
Hamilton® TLC syringes
1710, volume 100 μL, needle size 22 ga, needle L 51 mm (2 in.),
coated tip
Latch-lid TLC developing chambers
For use with 20 cm × 20 cm plates
Spectroline® E-Series UV lamp
output4 W, AC/DC input 115 V AC
Kimble® microcapillary pipettes
Kimble® microcapillary pipettes
Kimble® microcapillary pipettes
capacity 20 μL, accuracy: ±0.5%
Reagent sprayer for TLC plates
volume 250 mL, Bottle-Type
CONDITIONS
sample preparation
The boiled coffee samples were extracted and filtered with a 0.45 μm Millex syringe filter.
plate
HPTLC Silica gel 60 F254, 20 cm x 10 cm (1.05642.0001)
eluent
[A] acetonitrile; [B] methanol; [C] n-Heptane; (6:4:1, A:B:C)
migration distance
5 cm
sample
0.1 μL to 2.0 μL
detector
UV, 254 nm
Descrizione
Risultati analitici
Abstract: HPTLC (High Performance Thin Layer Chromatography) is a very useful analytical technique with very low sample preparation. Shown in the example: the quantification of caffeine in different coffee samples. The boiled coffee samples
were extracted and filtered with a 0.45 μm syringe filter. The plate was pre-conditioned with the mobile phase. After chromatographic separation the plate was scanned with a TLC-Scanner and subsequently quantified.
Results: Caffeine was detected under UV light (254 nm, hRf = 56, Fig. 1), (quantification information Fig. 2). A triple determination of each sample is possible in parallel.
Conclusion: These results demonstrate that caffeine can be easily quantified by TLC with a three-fold determination and minimal sample preparation.